Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM236 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583756
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583756 Supplier Invitrogen™ Supplier No. PA583756
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRGSQKSSEN GHIHSTSLQH IKTVTEQVRQ SPENAASPQA TNSTQVSQPS GAMTRSQESV FMGPQEPSCD SGILRMMSRR DVRAELFLWS FLLWSD Highest antigen sequence indentity to the following orthologs: Rat - 61%, Mouse - 60%.

TMEM236 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM236
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM236
Gene Accession No. Q5W0B7
Gene Alias bA162I21.2; bA16O1.2; FAM23A; FAM23B; family with sequence similarity 23, member A; family with sequence similarity 23, member B; TMEM236; Transmembrane protein 236
Gene Symbols TMEM236
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM236. Recombinant protein control fragment (Product #RP-108197).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653567
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.