Learn More
Invitrogen™ TMEM236 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 75%; Guinea Pig: 75%; Horse: 75%; Human: 100%.
Specifications
Specifications
| Antigen | TMEM236 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | TMEM236 |
| Gene Accession No. | Q5W0B7 |
| Gene Alias | bA162I21.2; bA16O1.2; FAM23A; FAM23B; family with sequence similarity 23, member A; family with sequence similarity 23, member B; TMEM236; Transmembrane protein 236 |
| Gene Symbols | TMEM236 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.