Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM237 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563013 Supplier Invitrogen™ Supplier No. PA563013
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NTPASASLEG LAQTAGRRPS EGNEPSTKEL KEHPEAPVQR RQKKTRLPLE LETSSTQKKS SSSSLLRNEN GIDA Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 65%.

The protein encoded by this gene is a tetraspanin protein that is thought to be involved in WNT signaling. Defects in this gene are a cause of Joubert syndrome-14. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jan 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM237
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM237
Gene Accession No. Q96Q45
Gene Alias AI853305; Als2cr4; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 4; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 4 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 4 protein homolog; Gm972; JBTS14; Tmem237; Transmembrane protein 237
Gene Symbols TMEM237
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM237. Recombinant protein control fragment (Product #RP-101391).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65062
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.