Learn More
Invitrogen™ TMEM24 Polyclonal Antibody

Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Specifications
Specifications
| Antigen | TMEM24 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | C2CD2L |
| Gene Accession No. | O14523 |
| Gene Alias | 1300006O23Rik; C2 calcium-dependent domain containing 2-like; C2 domain-containing protein 2-like; C2 domain-containing protein 2-like; phospholipid transfer protein C2CD2L; c2cd2 like; C2cd2l; C2CD2-like; DLNB23; KIAA0285; Phospholipid transfer protein C2CD2L; tmem24; Transmembrane protein 24; zgc:153961 |
| Gene Symbols | C2CD2L |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.