Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM240 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580139
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, NIH3T3 whole cell, COLO320 whole cell.
TMEM240 is a small, highly conserved transmembrane protein expressed in brain. The TMEM240 gene maps to chromosome 1p36.33. This gene encodes a transmembrane-domain containing protein found in the brain and cerebellum. Mutations in this gene result in spinocerebellar ataxia 21.
Specifications
TMEM240 | |
Polyclonal | |
Unconjugated | |
TMEM240 | |
C1orf70; EG381582; Gm5151; SCA21; Tmem240; Transmembrane protein 240; transmembrane protein C1orf70 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100362471, 339453, 381582 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
B2RWJ3, Q5SV17 | |
TMEM240 | |
E.coli-derived human TMEM240 recombinant protein (Position: M1-L173). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction