Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM243 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553256
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553256 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553256 Supplier Thermo Scientific Supplier No. PA553256
Add to Cart
Add to Cart

TMEM243 Polyclonal Antibody

TMEM243 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM243
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM243
Gene Accession No. Q9BU79
Gene Alias C7orf23; MDR1 and mitochondrial taxol resistance associated; MDR1- and mitochondrial taxol resistance-associated protein; MMTRAG; MM-TRAG; TMEM243; Transmembrane protein 243; transmembrane protein 243, mitochondrial; transmembrane protein C7orf23
Gene Symbols TMEM243
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM243. Recombinant protein control fragment (Product #RP-91102).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79161
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.