Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM248 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109297
Description
Positive Control: SkBr3, K562, SH-SY-5Y, HUVEC, SiHa.
TMEM248 is a protein coding gene.
Specifications
TMEM248 | |
Polyclonal | |
Unconjugated | |
TMEM248 | |
0610007L01Rik; A930023A16Rik; AW557951; C25H7orf42; C6H7orf42; C7orf42; G430067H08Rik; Tmem248; Transmembrane protein 248; Unknown (protein for MGC:134267); UPF0458 protein C7orf42; UPF0458 protein C7orf42 homolog; zgc:103561 | |
Rabbit | |
Affinity chromatography | |
RUO | |
55069 | |
-20°C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Flow Cytometry, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9NWD8 | |
TMEM248 | |
Synthetic peptide within Human Tmem248 aa 265-314. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction