Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM249 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140353
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: MPKGRAGSLP TTSIGWRFQL WFLGLTCPER HLARRLKNNS FYPFVQQEPN VFVLEYYL.
TMEM249 (Transmembrane Protein 249) is a Protein Coding gene and its function is unknown.
Specifications
| TMEM249 | |
| Polyclonal | |
| Unconjugated | |
| TMEM249 | |
| C8orfK29; EG666504; Gm8140; predicted gene 8140; putative transmembrane protein C8orfK29; putative transmembrane protein C8orfK29-like; TMEM249; Transmembrane protein 249 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 340393 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q2WGJ8 | |
| TMEM249 | |
| Recombinant protein corresponding to Human TMEM249. Recombinant protein control fragment (Product #RP-109687). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction