Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM25 Antibody (009), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290237AF750
Description
TMEM25 Monoclonal specifically detects TMEM25 in Human samples. It is validated for ELISA.Specifications
TMEM25 | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
84866 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
9 | |
ELISA | |
0610039J01Rik, FLJ14399, transmembrane protein 25 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human TMEM25 (AAH51841.1; Met1-Pro224). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction