Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM25 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300184JF646
Description
TMEM25 Polyclonal antibody specifically detects TMEM25 in Human samples. It is validated for ELISASpecifications
TMEM25 | |
Polyclonal | |
ELISA | |
0610039J01Rik, FLJ14399, transmembrane protein 25 | |
Produced in rabbits immunized with purified, recombinant Human TMEM25 (Accession#: AAH51841.1; Met1-Pro224) | |
0.1 mL | |
Cell Biology | |
84866 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction