Learn More
Invitrogen™ TMEM25 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589785
Description
Immunogen sequence: APGPSRHPSL ISSDSNNLKL NNVRLPRENM SLPSNLQLND LTPDSRAVKP ADRQMAQNNS RPELLDPEPG GLLTSQGFIR LPVLGYIYRV SSVSSDEIWL; Positive Samples: U-87MG, OVCAR3, LO2, Mouse brain, Mouse kidney, Rat brain, Rat kidney; Cellular Location: Cell membrane, Single-pass type I membrane protein.
TMEM25 (Transmembrane Protein 25) is a Protein Coding gene. An important paralog of this gene is SIGLEC1.
Specifications
TMEM25 | |
Polyclonal | |
Unconjugated | |
TMEM25 | |
0610039J01Rik; AI429491; Tmem25; Transmembrane protein 25; UNQ2531/PRO6030 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
689172, 71687, 84866 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.09 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q86YD3, Q9DCF1 | |
TMEM25 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 223-322 of human TMEM25 (NP_0011375061). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.