Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM25 Rabbit anti-Human, Clone: 9, Novus Biologicals™
SDP

Catalog No. p-7194506 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021938 50 μg
NB021937 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021938 Supplier Novus Biologicals Supplier No. NBP29023750UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TMEM25 Monoclonal specifically detects TMEM25 in Human samples. It is validated for ELISA.

Specifications

Antigen TMEM25
Applications ELISA
Classification Monoclonal
Clone 9
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias 0610039J01Rik, FLJ14399, transmembrane protein 25
Host Species Rabbit
Immunogen Recombinant Human TMEM25 Protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84866
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.