Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM253 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562587
Description
Immunogen sequence: LLSQRKPGCC RSQSLHYQEL QEGFSELEEV PGLENGPTVA STGANERVGQ REQTRAALLP P Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 51%.
Specifications
| TMEM253 | |
| Polyclonal | |
| Unconjugated | |
| TMEM253 | |
| C14orf176; C14orf95; NCRNA00220; TMEM253; Transmembrane protein 253; transmembrane protein C14orf176 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 643382 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P0C7T8 | |
| TMEM253 | |
| Recombinant protein corresponding to Human TMEM253. Recombinant protein control fragment (Product #RP-100568). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction