Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM254 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567633
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567633 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567633 Supplier Invitrogen™ Supplier No. PA567633
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM254 Polyclonal Antibody for Western Blot

TMEM254 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM254
Applications Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TMEM254
Gene Accession No. P0DN89, Q8TBM7
Gene Alias 0610008K04Rik; 0610010A22Rik; AA960404; bA369J21.6; C10orf57; D14Ertd449e; Tmem254; Tmem254a; Tmem254b; Tmem254c; tranmembrane protein 254a; transmembrane protein 254; transmembrane protein 254a; Transmembrane protein 254b; Transmembrane protein 254c; transmembrane protein C10orf57; transmembrane protein C10orf57 homolog
Gene Symbols TMEM254, Tmem254a
Host Species Rabbit
Immunogen Synthesized peptide derived from human TMEM254.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66039, 80195
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.