Learn More
Invitrogen™ TMEM254 Polyclonal Antibody

Description
Immunogen sequence: FWPQSIPYQN LGPLGPFTQY LVDHHHTL Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 86%.
Specifications
Specifications
| Antigen | TMEM254 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TMEM254 |
| Gene Accession No. | Q8TBM7 |
| Gene Alias | 0610008K04Rik; 0610010A22Rik; AA960404; bA369J21.6; C10orf57; D14Ertd449e; Tmem254; Tmem254a; Tmem254b; Tmem254c; tranmembrane protein 254a; transmembrane protein 254; transmembrane protein 254a; Transmembrane protein 254b; Transmembrane protein 254c; transmembrane protein C10orf57; transmembrane protein C10orf57 homolog |
| Gene Symbols | TMEM254 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.