Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM255B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144063
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144063 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144063 Supplier Invitrogen™ Supplier No. PA5144063
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: mouse heart tissue, mouse testis tissue, mouse lung tissue. IHC: rat lung tissue, rat intestine tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The TMEM255B gene encodes a member of the transmembrane protein family, which are proteins that span biological membranes and have been suggested to have roles in cellular signaling and structure. TMEM255B has been studied for its expression in different cancer types, where it acts as a potential marker. Specifically, it has been observed in serous cystic neoplasms (SCNs) and other neoplasms, with a noteworthy frequency in gastric-type intraductal papillary mucinous neoplasms (IPMNs) and pancreatic ductal adenocarcinomas (PDACs). The expression of TMEM255B in these neoplasms indicates its potential as a diagnostic marker and may reflect its involvement in cancer biology. While its functions are still being elucidated, TMEM255B's role in cancer highlights its significance in tumor progression and potential as a target for further investigation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM255B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.01mg sodium azide
Gene TMEM255B
Gene Accession No. Q5FW56
Gene Alias FAM70B; family with sequence similarity 70, member B; protein FAM70B; TMEM255B; Transmembrane protein 255B
Gene Symbols TMEM255B
Host Species Rabbit
Immunogen E.coli-derived mouse Tmem255b recombinant protein (Position: Q196-L240).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 272465, 290877
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.