Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM258 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560772
Description
Immunogen sequence: MELEAMSRYT SPVNPAVFPH LTVVL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Specifications
TMEM258 | |
Polyclonal | |
Unconjugated | |
TMEM258 | |
C11orf10; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit TMEM258; HSPC005; Oligosaccharyl transferase subunit TMEM258; TMEM258; Transmembrane protein 258; UPF0197 transmembrane protein C11orf10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
746 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P61165 | |
TMEM258 | |
Recombinant protein corresponding to Human TMEM258. Recombinant protein control fragment (Product #RP-97349). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction