Learn More
Description
Specifications
Specifications
| Antigen | TMEM26 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 - 2 ug/mL, Simple Western 1:20, Flow Cytometry, Immunohistochemistry reported in scientific literature (PMID 30089732), Immunohistochemistry-Paraffin reported in scientific literature (PMID 28250021) |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Accession No. | Q6ZUK4 |
| Gene Alias | DKFZp686D09128, Em:AC068892.1, MGC35010, transmembrane protein 26 |
| Gene Symbols | TMEM26 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
