Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM260 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568631
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 92%; Rabbit: 86%; Rat: 93%.
TMEM260 is a protein coding gene.
Specifications
TMEM260 | |
Polyclonal | |
Unconjugated | |
TMEM260 | |
5930435P15; 6720456H20Rik; C10H14orf101; C14orf101; C1H14orf101; C8H14orf101; LOW QUALITY PROTEIN: transmembrane protein 260; RGD1562018; TMEM260; Transmembrane protein 260; UPF0679 protein C14orf101; UPF0679 protein C14orf101 homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54916 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NX78 | |
TMEM260 | |
synthetic peptide directed towards the N terminal region of human TMEM260. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction