Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM260 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568632
Description
This target displays homology in the following species: Cow: 79%; Dog: 86%; Horse: 86%; Human: 100%; Mouse: 86%; Pig: 86%; Rabbit: 86%; Rat: 86%; Yeast: 75%.
TMEM260 is a protein coding gene.
Specifications
TMEM260 | |
Polyclonal | |
Unconjugated | |
TMEM260 | |
5930435P15; 6720456H20Rik; C10H14orf101; C14orf101; C1H14orf101; C8H14orf101; LOW QUALITY PROTEIN: transmembrane protein 260; RGD1562018; TMEM260; Transmembrane protein 260; UPF0679 protein C14orf101; UPF0679 protein C14orf101 homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
218989 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8BMD6 | |
TMEM260 | |
synthetic peptide directed towards the middle region of mouse TMEM260. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction