Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM261 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553197
Description
Immunogen sequence: MGSRLSQPFE SYITAPPGTA AAPAKPAPPA TPGAPTSPAE HRLLKTCWSC Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 41%.
Required for the assembly of the mitochondrial NADH:ubiquinone oxidoreductase complex (complex I). Involved in the assembly of the distal region of complex I. [UniProt].
Specifications
| TMEM261 | |
| Polyclonal | |
| Unconjugated | |
| DMAC1 | |
| 1700027K24Rik; 3110001D03Rik; C9orf123; distal membrane arm assembly complex 1; Distal membrane-arm assembly complex protein 1; Dmac1; Tmem261; Transmembrane protein 261; transmembrane protein C9orf123; transmembrane protein C9orf123 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90871 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96GE9 | |
| DMAC1 | |
| Recombinant protein corresponding to Human TMEM261. Recombinant protein control fragment (Product #RP-90965). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction