Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM261 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA546291
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546291 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546291 Supplier Thermo Scientific Supplier No. PA546291

Rabbit Polyclonal Antibody

Peptide sequence: MGAGGYVYWV ARKPMKMGYP PSPWTITQMV IGLSIATWGI VVMADPKGKA Sequence homology: Human: 100%; Mouse: 75%; Rabbit: 85%

TMEM261 Polyclonal antibody specifically detects TMEM261 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM261
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DMAC1
Gene Accession No. Q96GE9
Gene Alias 1700027K24Rik; 3110001D03Rik; C9orf123; distal membrane arm assembly complex 1; Distal membrane-arm assembly complex protein 1; Dmac1; Tmem261; Transmembrane protein 261; transmembrane protein C9orf123; transmembrane protein C9orf123 homolog
Gene Symbols DMAC1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human C9orf123
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90871
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.