Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM27 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561766
Description
Immunogen sequence: LVTAIHAELC QPGAENAFKV RLSIRTALGD KAYAWDTNEE YLFKAMVAFS MRKVPNREAT EISHVLLCNV TQRVSFWFVV TDPSKNHTLP AVEVQSAIRM NKNRINNAFF LNDQTLE Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.
TMEM27 (Transmembrane protein 27) is also named as collectrin and Belongs to the TMEM27 family. It is a stimulator of beta cell replication and a membrane protein cleaved and shed by pancreatic beta cells that has been proposed as a beta cell mass biomarker. Western blot analysis of mouse collecting duct cell lines detected Tmem27 at an apparent molecular mass of 32 kDa. TMEM27 is an N-glycosylated protein and forms dimers, with MW of 64-75 kDa.
Specifications
TMEM27 | |
Polyclonal | |
Unconjugated | |
TMEM27 | |
0610008J07Rik; Cltrn; Collectrin; kidney-specific membrane protein; NX17; NX-17; Tmem27; Transmembrane protein 27; UNQ679/PRO1312 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57393 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HBJ8 | |
TMEM27 | |
Recombinant protein corresponding to Human TMEM27. Recombinant protein control fragment (Product #RP-91896). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction