Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM27 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588721
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588721 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588721 Supplier Invitrogen™ Supplier No. PA588721
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLWLLFFLVT AIHAELCQPG AENAFKVRLS IRTALGDKAY AWDTNEEYLF KAMVAFSMRK VPNREATEIS HVLLCNVTQR VSFWFVVTDP SKNHTLPAVE VQSAIRMNKN RINNAFFLND QTLEFLKIPS TLAPPMDPSV PIWIIIFGVI FCIIIVAIAL LILSGIWQRR RKNKEPSEVD DAEDKCENMI TIENGIPSDP LDMKGGHIND AFMTEDERLT PL; Positive Samples: Mouse Kidney, Mouse Pancreas, Rat Kidney; Cellular Location: Membrane, Single-pass type I membrane protein.

TMEM27 (Transmembrane protein 27) is also named as collectrin and Belongs to the TMEM27 family. It is a stimulator of beta cell replication and a membrane protein cleaved and shed by pancreatic beta cells that has been proposed as a beta cell mass biomarker. Western blot analysis of mouse collecting duct cell lines detected Tmem27 at an apparent molecular mass of 32 kDa. TMEM27 is an N-glycosylated protein and forms dimers, with MW of 64-75 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM27
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TMEM27
Gene Accession No. Q9ESG3, Q9ESG4
Gene Alias 0610008J07Rik; Cltrn; Collectrin; kidney-specific membrane protein; NX17; NX-17; Tmem27; Transmembrane protein 27; UNQ679/PRO1312
Gene Symbols TMEM27
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-222 of human TMEM27 (NP_0657161).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57394, 57395
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.