Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM27 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | TMEM27 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4223
![]() ![]() |
Proteintech Group Inc
240591AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
TMEM27 (Transmembrane protein 27) is also named as collectrin and belongs to the TMEM27 family. It is a pancreatic beta-cell surface protein promoting the preservation of functional beta-cell mass and may be responsible for the sodium retention in salt-sensitive hypertension. This full length has two glycosylation sites.Specifications
TMEM27 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9HBJ8 | |
57393 | |
TMEM27 Fusion Protein Ag21257 | |
Primary | |
-20°C | |
TMEM27 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Collectrin, NX 17, NX17, TMEM27, transmembrane protein 27 | |
TMEM27 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title