Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM30B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560263
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560263 Supplier Invitrogen™ Supplier No. PA560263
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GIKELEYDYT GDPGTGNCSV CAAAGQGRAL PPPCSCAWYF SLPELFQGPV YLYYELTNFY QNNRRYGVSR DDAQLSGLPS ALRHPVNEC Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.

Accessory component of a P4-ATPase flippase complex which catalyzes the hydrolysis of ATP coupled to the transport of aminophospholipids from the outer to the inner leaflet of various membranes and ensures the maintenance of asymmetric distribution of phospholipids. Phospholipid translocation seems also to be implicated in vesicle formation and in uptake of lipid signaling molecules. The beta subunit may assist in binding of the phospholipid substrate (Probable). Can mediate the export of alpha subunits ATP8A1, ATP8B1, ATP8B2 and ATP8B4 from the ER to the plasma membrane. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM30B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM30B
Gene Accession No. Q3MIR4
Gene Alias 9130011B11Rik; CDC50B; Cell cycle control protein 50B; P4-ATPase flippase complex beta subunit TMEM30B; Tmem30b; Transmembrane protein 30B
Gene Symbols TMEM30B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM30B. Recombinant protein control fragment (Product #RP-99945).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161291
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.