Learn More
Invitrogen™ TMEM30B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113797
Description
Immunogen sequence: VYLYYELTNF YQNNRRYGVS RDDAQLSGLP SALRHPVNEC APYQRSAAGL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Accessory component of a P4-ATPase flippase complex which catalyzes the hydrolysis of ATP coupled to the transport of aminophospholipids from the outer to the inner leaflet of various membranes and ensures the maintenance of asymmetric distribution of phospholipids. Phospholipid translocation seems also to be implicated in vesicle formation and in uptake of lipid signaling molecules. The beta subunit may assist in binding of the phospholipid substrate (Probable). Can mediate the export of alpha subunits ATP8A1, ATP8B1, ATP8B2 and ATP8B4 from the ER to the plasma membrane. [UniProt].
Specifications
| TMEM30B | |
| Polyclonal | |
| Unconjugated | |
| TMEM30B | |
| 9130011B11Rik; CDC50B; Cell cycle control protein 50B; P4-ATPase flippase complex beta subunit TMEM30B; Tmem30b; Transmembrane protein 30B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 161291 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q3MIR4 | |
| TMEM30B | |
| Synthetic peptide directed towards the middle region of human TMEM30B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.