Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144111
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144111 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144111 Supplier Invitrogen™ Supplier No. PA5144111
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM31 Polyclonal Antibody for ICC/IF, IHC (P), ELISA

TMEM31 is a gene that encodes a transmembrane protein with unknown function. It is located on the long arm of chromosome 5 in humans and is conserved across different species. While the specific function of TMEM31 is not well understood, studies have suggested that it may be involved in the regulation of autophagy, a process by which cells break down and recycle their own components.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM31
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene TMEM31
Gene Accession No. Q5JXX7
Gene Alias TMEM31; Transmembrane protein 31
Gene Symbols TMEM31
Host Species Rabbit
Immunogen Recombinant Human Transmembrane protein 31 protein (1-118AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203562
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.