Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM33 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569469
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569469 Supplier Invitrogen™ Supplier No. PA569469
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 100%; Zebrafish: 92%.

Acts as a regulator of the tubular endoplasmic reticulum (ER) network. Suppresses the RTN3/4-induced formation of the ER tubules. Positively regulates PERK-mediated and IRE1-mediated unfolded protein response signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM33
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMEM33
Gene Accession No. Q9Z142
Gene Alias 1110006G02Rik; 1600019D15Rik; 2410089A21Rik; 2700052H22Rik; 5430406L04Rik; AA388285; AI314185; AI426615; db83; hypothetical protein LOC327214; Protein DB83; SHINC3; SHINC-3; Tmem33; Transmembrane protein 33; wu:fd16h10; wu:fi11f08; zgc:55931; zgc:77670
Gene Symbols TMEM33
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of rat TMEM33 (aa 120-169).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59303
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.