Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM37 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116053
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116053 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116053 Supplier Invitrogen™ Supplier No. PA5116053
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TMEM37.

Thought to stabilize the calcium channel in an inactivated (closed) state. Modulates calcium current when coexpressed with CACNA1G.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM37
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene TMEM37
Gene Accession No. Q8VHW1, Q8WXS4, Q9JJV3
Gene Alias AI173373; AW540203; Cacng5; Neuronal voltage-gated calcium channel gamma-like subunit; Pr; Pr1; protein distantly related to to the gamma subunit family; TMEM37; tmem37 {ECO:0000250; Transmembrane protein 37; UniProtKB:Q9JJV3}; voltage-dependent calcium channel gamma subunit-like protein; voltage-dependent calcium channel gamma-like subunit
Gene Symbols TMEM37
Host Species Rabbit
Immunogen A synthesized peptide derived from human TMEM37(Accession Q8WXS4), corresponding to amino acid residues M1-S42.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140738, 170706, 245953
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.