Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM37 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562842
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562842 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562842 Supplier Invitrogen™ Supplier No. PA562842
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LMVSQLCEDK HSQCKWVMGS Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 60%.

Thought to stabilize the calcium channel in an inactivated (closed) state. Modulates calcium current when coexpressed with CACNA1G.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM37
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM37
Gene Accession No. Q8WXS4
Gene Alias AI173373; AW540203; Cacng5; Neuronal voltage-gated calcium channel gamma-like subunit; Pr; Pr1; protein distantly related to to the gamma subunit family; TMEM37; tmem37 {ECO:0000250; Transmembrane protein 37; UniProtKB:Q9JJV3}; voltage-dependent calcium channel gamma subunit-like protein; voltage-dependent calcium channel gamma-like subunit
Gene Symbols TMEM37
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM37. Recombinant protein control fragment (Product #RP-99899).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140738
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.