Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM38A Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297578MFV500
Description
TMEM38A Polyclonal antibody specifically detects TMEM38A in Human, Cynomolgus Monkey samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
TMEM38A | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
transmembrane protein 38ATRIC-AMGC3169, TRICA, trimeric intracellular cation channel type A | |
Produced in rabbits immunized with E. coli-derived Human TMEM38A fragment. | |
0.1 mL | |
Signal Transduction | |
79041 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human, Cynomolgus Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction