Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM38B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553809
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553809 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553809 Supplier Invitrogen™ Supplier No. PA553809
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FEDTLSWMLF GWQQPFSSCE KKSEAKSPSN GVGSLASKPV DVASDNV Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 52%.

TMEM38A and TMEM38B are two recently identified trimeric intracellular cation (TRIC) channel subtypes. TMEM38B is expressed in most mammalian tissues, while TMEM38A is preferentially expressed in excitable tissues such as striated muscle and brain. Mice deficient in both TMEM38A and TMEM38B suffer embryonic cardiac failure; the cardiac myocytes display severe dysfunction in SR Ca2+ handling, weakened Ca2+ release, and reduced K+ permeability indicating that the TRIC cation channels are likely to act as counter-ion channels that function in synchronization with Ca2+ release from intracellular stores. Mice that were lacking only TMEM38B however, die shortly after birth due to respiratory failure and have lungs exhibiting severe histological defect and ultrastructural abnormalities in their alveolar type II epithelial cells, indicating that TMEM38B are essential for perinatal lung maturation. Other experiments have shown that TMEM38A and TMEM38B can act with junctophilin proteins to support efficient ryanodine receptor-mediated Ca2+ release in muscle cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM38B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM38B
Gene Accession No. Q9NVV0
Gene Alias 1600017F22Rik; AA437809; AV051057; bA219P18.1; C9orf87; D4Ertd89e; mg33b; Mitsugumin33B; Mitsugumin-33B; OI14; RGD1305703; RP11-219P18.1; TMEM38B; Transmembrane protein 38B; TRICB; TRIC-B; trimeric intracellular cation channel type B
Gene Symbols TMEM38B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM38B. Recombinant protein control fragment (Product #RP-91197).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55151
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.