Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM39A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586009
Description
TMEM39A Polyclonal Antibody for ICC/IF, IHC (P), Flow
TMEM39A is a protein coding gene.
Specifications
TMEM39A | |
Polyclonal | |
Unconjugated | |
TMEM39A | |
2610033C09Rik; RGD1306421; TMEM39A; Transmembrane protein 39A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55254, 67846 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
Q9CYC3, Q9NV64 | |
TMEM39A | |
Synthetic peptide corresponding to Human TMEM39a aa 475-488. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction