Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM41B Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP312857MFV450
Description
TMEM41B Polyclonal antibody specifically detects TMEM41B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
TMEM41B | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
KIAA0033MGC33897, MGC57262, transmembrane protein 41B | |
Anti-TMEM41B antibody was raised against a peptide corresponding to 10 amino acids near the carboxyl terminus of human TMEM41B. The immunogen is located within the last 50 amino acids of TMEM41B. | |
0.1 mL | |
Autophagy, Lipid and Metabolism | |
440026 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction