Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM41B Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300527MFV450
Description
TMEM41B Polyclonal antibody specifically detects TMEM41B in Human, Mouse samples. It is validated for Western BlotSpecifications
TMEM41B | |
Polyclonal | |
Western Blot | |
KIAA0033MGC33897, MGC57262, transmembrane protein 41B | |
Partial synthetic peptide made to an internal portion of the human TMEM41B protein (between amino acids 50-100) [UniProt Q5BJD5]. | |
0.1 mL | |
Autophagy, Lipid and Metabolism | |
440026 | |
Store at 4C in the dark. |
Western Blot | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction