Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM41B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143836
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143836 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143836 Supplier Invitrogen™ Supplier No. PA5143836
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted species reactivity based on immunogen sequence: Bovine (100%); Chicken (100%); Chimpanzee (100%).

TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM41B
Applications Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TMEM41B
Gene Accession No. Q5BJD5, Q5FVN2, Q8K1A5
Gene Alias KIAA0033; TMEM41B; Transmembrane protein 41B
Gene Symbols TMEM41B
Host Species Rabbit
Immunogen A peptide corresponding to 10 amino acids near the carboxyl terminus of human TMEM41B. The immunogen is located within the last 50 amino acids of TMEM41B.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233724, 361626, 440026
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.