Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM43 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305860PE
Description
TMEM43 Polyclonal antibody specifically detects TMEM43 in Human samples. It is validated for Western BlotSpecifications
TMEM43 | |
Polyclonal | |
Western Blot | |
ARVC5, ARVD5, DKFZp586G1919, LUMA, MGC3222, Protein LUMA, transmembrane protein 43 | |
Produced in rabbits immunized with E. coli-derived Human TMEM43 fragment. | |
0.1 mL | |
Signal Transduction | |
79188 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction