Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM44 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110498
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110498 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110498 Supplier Invitrogen™ Supplier No. PA5110498
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VCGSKFKSNS DREARERKRR RQLRASVFAL ALPLSLGPCW ALWVAVPKAS ATIRGPQRRL LASLLQENTE ILGYLLGSVA AFGSWASRIP PLSRIAPPPT LGITTQHEIW RGQMSKPSQS PSRSPSGHWR AAAQRQVLGT EMCRGKTFPS I.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM44
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TMEM44
Gene Accession No. Q2T9K0
Gene Alias 1700007N03Rik; 9330161C17Rik; B230220N21; RGD1563737; TMEM44; Transmembrane protein 44
Gene Symbols TMEM44
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 110-260 of human TMEM44 (NP_001159777.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93109
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.