Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM45A Polyclonal Antibody, Invitrogen™

Catalog No. PIPA564003
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564003 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564003 Supplier Thermo Scientific Supplier No. PA564003

Rabbit Polyclonal Antibody

Immunogen sequence: TKSILKYICK KQKRTCYLGS KTLFYRLEI Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 66%.

TMEM45A Polyclonal antibody specifically detects TMEM45A in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM45A
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM45A
Gene Accession No. Q9NWC5
Gene Alias dermal papilla derived protein 7; dermal papilla-derived protein 7; DERP7; DNA polymerase-transactivated protein 4; DNAPTP4; TMEM45A; Transmembrane protein 45A
Gene Symbols TMEM45A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM45A
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55076
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.