Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM48 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542844
Description
Peptide sequence: SFTEDRFGVV QTTLPAILNT LLTLQEAVDK YFKLPHASSK PPRISGSLVD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
TMEM48 is a component of the nuclear pore complex (NPC), which plays a key role in de novo assembly and insertion of NPC in the nuclear envelope. TMEM48 is required for NPC and nuclear envelope assembly, possibly by forming a link between the nuclear envelope membrane and soluble nucleoporins, thereby anchoring the NPC in the membrane.
Specifications
TMEM48 | |
Polyclonal | |
Unconjugated | |
NDC1 | |
2810475A17Rik; AI450313; FLJ10407; FLJ12556; FLJ34120; hNDC1; ndc1; NDC1 transmembrane nucleoporin; NET3; nuclear division cycle 1 homolog; nucleoporin NDC1; RP4-654H19.1; tmem48; Transmembrane protein 48; wu:fk93f04; zgc:55636 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55706 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BTX1 | |
NDC1 | |
Synthetic peptide directed towards the middle region of human TMEM48. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction