Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM49 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987462
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7462 Supplier Bioss Supplier No. BS6740R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VMP1 is a stress-induced protein that, when overexpressed, promotes formation of intracellular vacuoles followed by cell death. May be involved in the cytoplasmic vacuolization of acinar cells during the early stage of acute pancreatitis.VMP1 is a transmembrane protein that co-localizes with LC3, a marker of the autophagosomes. It interacts with beclin-1, a mammalian autophagy initiator, through the VMP1-Atg domain, which is essential for autophagosome formation.

Specifications

Antigen TMEM49
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene VMP1
Gene Accession No. Q96GC9
Gene Alias 3110098I04Rik; 4930579A11Rik; AI787464; BOS_18345; EPG3; HSPC292; LOC100351532; mir-21a; NF-E2 induceble protein-2; NF-E2-inducible protein 2; ni-2; Protein ni-2; TANGO5; TDC1; tmem49; Transmembrane protein 49; Vacuole membrane protein 1; vmp1; wu:fb05d08; wu:fb92g08; zgc:77153
Gene Symbols VMP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TMEM49.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81671
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.