Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566657
Description
Immunogen sequence: SEEWNPWEGDE KNEQQHRFKT SLQILDKSTK GKTDLSVQIW GKAAIGLYLW EHIFEGLLDP SDVTAQWREG KSIVGRTQ Highest antigen sequence identity to the following orthologs - mouse 82%, rat 81%.
Involved in the biosynthesis of the phosphorylated O-mannosyl trisaccharide (N-acetylgalactosamine-beta-3-N-acetylglucosamine-beta-4-(phosphate-6-)mannose), a carbohydrate structure present in alpha-dystroglycan (DAG1), which is required for binding laminin G-like domain-containing extracellular proteins with high affinity.
Specifications
TMEM5 | |
Polyclonal | |
Unconjugated | |
RXYLT1 | |
6330415D21Rik; HP10481; MDDGA10; Ribitol-5-phosphate xylosyltransferase 1; RXYLT1; TMEM5; Transmembrane protein 5; type II membrane protein; UDP-D-xylose:ribitol-5-phosphate beta1,4-xylosyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10329 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2B1 | |
RXYLT1 | |
Recombinant Human TMEM5. Recombinant protein control fragment (Product #RP-105298). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction