Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM50B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567028
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA567028 100 μL
1 options

Catalog No. PIPA567028

Supplier: Invitrogen™ PA567028

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DAAVVYPKPEQ LNHAFHTCG Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

TMEM50B gene ontology annotations related to this gene include late endosome to vacuole transport via multivesicular body sorting pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM50B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM50B
Gene Accession No. P56557
Gene Alias AU015466; AU019872; B230114J08Rik; C21orf4; HCV p7-transregulated protein 3; HCV p7-trans-regulated protein 3; HCVP7TP3; RGD1307561; TMEM50B; Transmembrane protein 50B; transmembrane protein 50B-like protein; UNQ167/PRO193
Gene Symbols TMEM50B
Host Species Rabbit
Immunogen Recombinant Human TMEM50B. Recombinant protein control fragment (Product #RP-107667).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 757
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.