Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM50B Polyclonal Antibody

TMEM50B Polyclonal Antibody
Supplier: Thermo Scientific PA570468
Description
TMEM50B Polyclonal Antibody for Western Blot
Specifications
| TMEM50B | |
| Polyclonal | |
| Unconjugated | |
| TMEM50B | |
| AU015466; AU019872; B230114J08Rik; C21orf4; HCV p7-transregulated protein 3; HCV p7-trans-regulated protein 3; HCVP7TP3; RGD1307561; TMEM50B; Transmembrane protein 50B; transmembrane protein 50B-like protein; UNQ167/PRO193 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 77975 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9D1X9 | |
| TMEM50B | |
| synthetic peptide directed towards the middle region of human TMEM50B (aa 49-98). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction