Learn More
Invitrogen™ TMEM51 Polyclonal Antibody

Description
Immunogen sequence: EDSQEEEEEDE EAASRYYVPS YEEVMNTNYS EARGEEQNPR LSISLPSYES LTGLDETTPT STRADVEASP GNPPDRQNSK LAKRLKPLKV RRIKSEKLHL KDFRITLP Highest antigen sequence identity to the following orthologs - mouse 85%, rat 86%.
Specifications
Specifications
| Antigen | TMEM51 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TMEM51 |
| Gene Accession No. | Q9NW97 |
| Gene Alias | C1orf72; TMEM51; Transmembrane protein 51 |
| Gene Symbols | TMEM51 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.