Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM52 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145210
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145210 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145210 Supplier Invitrogen™ Supplier No. PA5145210
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VIPMDSDSPV HSTVTSYSSV QYPLGMRLPL PFGELDLDSV APPAYSLYTP EPPPSYDEAV KMAKPREEGP ALSQKPSPLL GASGLETTPV PQESGPNTQL PP Antigen sequence identity: Mouse 68%; Rat 70%.

TMEM52 is a transmembrane protein that is involved in the regulation of autophagy, a process by which cells break down and recycle their own components. It is also thought to play a role in the development of certain types of cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM52
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM52
Gene Accession No. Q8NDY8
Gene Symbols TMEM52
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 102-203.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339456
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.