Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM52B Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298159APC
Description
TMEM52B Polyclonal antibody specifically detects TMEM52B in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
TMEM52B | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
C12orf59, chromosome 12 open reading frame 59, UNQ5927/PRO19821 | |
Produced in rabbits immunized with E. coli-derived Human TMEM52B fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
120939 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction