Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM55A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110499
Description
Immunogen sequence: VMLISEEQPA QPALPIQPEG TRVVCGHCGN TFLWMELRFN TLAKCPHCKK ISSVGSALPR RRCCAY.
TMEM55A (PIP4P2, Phosphatidylinositol 4,5 Bisphosphate 4 Phosphatase 2) is a Protein Coding gene. Among its related pathways aresuperpathway of inositol phosphate compounds. An important paralog of this gene isPIP4P1.
Specifications
TMEM55A | |
Polyclonal | |
Unconjugated | |
PIP4P2 | |
PIP4P2; PtdIns-4,5-P(2) 4-phosphatase type II; ptdIns-4,5-P2 4-Ptase II; TMEM55A; Transmembrane protein 55A; type 2 phosphatidylinositol 4,5-bisphosphate 4-phosphatase; Type 2 PtdIns-4,5-P2 4-Ptase; type II phosphatidylinositol 4,5-bisphosphate 4-phosphatase; type II phosphatidylinositol-4,5-bisphosphate 4-phosphatase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55529 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.15 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8N4L2 | |
PIP4P2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 130-195 of human TMEM55A (NP_061180.1). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction