Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM55A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112760
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112760 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112760 Supplier Invitrogen™ Supplier No. PA5112760
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM55A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

TMEM55A (PIP4P2, Phosphatidylinositol 4,5 Bisphosphate 4 Phosphatase 2) is a Protein Coding gene. Among its related pathways aresuperpathway of inositol phosphate compounds. An important paralog of this gene isPIP4P1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM55A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PIP4P2
Gene Accession No. Q8N4L2
Gene Alias PIP4P2; PtdIns-4,5-P(2) 4-phosphatase type II; ptdIns-4,5-P2 4-Ptase II; TMEM55A; Transmembrane protein 55A; type 2 phosphatidylinositol 4,5-bisphosphate 4-phosphatase; Type 2 PtdIns-4,5-P2 4-Ptase; type II phosphatidylinositol 4,5-bisphosphate 4-phosphatase; type II phosphatidylinositol-4,5-bisphosphate 4-phosphatase
Gene Symbols PIP4P2
Host Species Rabbit
Immunogen Recombinant Human Type 2 phosphatidylinositol 4,5-bisphosphate 4-phosphatase protein (1-191AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55529
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.